missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrachloropalladate(II), Premion™, 99.998% (metals basis), Pd 37%

Product Code. 11356957
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11356957

Brand: Thermo Scientific Alfa Aesar 010824.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13820-40-1 | Cl4H8N2Pd | 284.30 g/mol

Ammonium tetrachloropalladate(II) is one of the preferred complexes, which are used for the preparation of palladium metallic particles. In addition, it forms Pd-creatinine complexes, which find use in the field of catalysis, and semiconductors. Colloidal palladium nanocatalysts and metal polymer catalyst systems can be prepared from ammonium tetrachloropalladate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-40-1
Molecular Formula Cl4H8N2Pd
Molecular Weight (g/mol) 284.30
MDL Number MFCD00010884
InChI Key IRKVTUVFHGUMMN-UHFFFAOYSA-L
Synonym ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride
PubChem CID 61682
ChEBI CHEBI:60149
IUPAC Name diazanium;tetrachloropalladium(2-)
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++]

Specifications

Melting Point (decomposition)
Color Brown to Green
Physical Form Powder
Quantity 5 g
Linear Formula (NH4)2PdCl4
Solubility Information Soluble in water
Formula Weight 284.31
Percent Purity 99.998%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 37%
Sensitivity Hygroscopic
Density 2.17 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium tetrachloropalladate(II)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.