Learn More
Labconco™ Ampules
Shop All Labconco Corporation Products
Click to view available options
Capacity (Metric):
1 mL
5 mL
10 mL
Description:
Pre-scored, flat-bottom
Straight-sided
Unit Size:
100 ampoules
Pack of 100
Description
- Constructed of high quality borosilicate glass for strength and durability
- Supplied in packages of 100
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Material | Borosilicate Glass |
| Product Type | Ampule |
| Description | Pre-scored, flat-bottom |