missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amylase alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
141.00 EUR - 413.00 EUR
Specifications
Antigen | Amylase alpha |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17793644
|
Invitrogen
PA5122251 |
20 μL |
141.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17783644
|
Invitrogen
PA5122250 |
100 μL |
413.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.Specifications
Amylase alpha | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
P00689, P04746, P19961 | |
279, 280, 497039 | |
Asp188-Val432 (Accession P04746), with N-terminal His Tag | |
Primary | |
-20°C | |
AMY2B |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; a-amylase; alpha-amylase; Alpha-amylase 2B; AMY2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a3; Amy2b; AMY3; amylase 2, pancreatic; amylase 2-2, pancreatic; amylase 2a1; amylase 2a3; amylase 2a3, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; mAmy2-4; mAmy2-6; OTTMUSG00000022430; OTTMUSG00000022432; OTTMUSG00000022462; PA; pancreatic alpha-amylase; pancreatic amylase; pancreatic amylase alpha 2A | |
AMY2A, Amy2a3, AMY2B | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title