missing translation for 'onlineSavingsMsg'
Learn More

Anacardic acid, Tocris Bioscience™

Product Code. 18004742
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Noncompetitive PCAF/p300 inhibitor

Noncompetitive inhibitor of PCAF and p300 histone acetyltransferase (HAT) activity (IC50 values are ∽5.0 and ∽8.5 μM respectively). Inhibits NF-κB activation. Also reported to selectively activate Aurora kinase A-mediated phosphorylation of histone H3 and to inhibit protein SUMOylation. Exhibits antitumor, antimicrobial and antioxidant activity.

Chemical Identifiers

CAS 16611-84-0
Molecular Formula C22H36O3
Molecular Weight (g/mol) 348.527
InChI Key ADFWQBGTDJIESE-UHFFFAOYSA-N
Synonym anacardic acid, 6-pentadecylsalicylic acid, cyclogallipharic acid, 22:0-anacardic acid, 2-hydroxy-6-pentadecyl-benzoic acid, ginkgolic acid c15:0, anacardsaure, anarcadic acid, anacardic acid a, anacardic acid /
PubChem CID 167551
ChEBI CHEBI:2696
IUPAC Name 2-hydroxy-6-pentadecylbenzoic acid
SMILES CCCCCCCCCCCCCCCC1=C(C(=CC=C1)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 348.52
Percent Purity >97%
Chemical Name or Material Anacardic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.