missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anacardic acid, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Noncompetitive inhibitor of PCAF and p300 histone acetyltransferase (HAT) activity (IC50 values are ∽5.0 and ∽8.5 μM respectively). Inhibits NF-κB activation. Also reported to selectively activate Aurora kinase A-mediated phosphorylation of histone H3 and to inhibit protein SUMOylation. Exhibits antitumor, antimicrobial and antioxidant activity.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 348.52 |
Percent Purity | >97% |
Chemical Name or Material | Anacardic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction