Learn More
BINDER™ Analog Output
Product Code. 30238335 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65/130/260/470/720/1060
KB PRO 130/260/470
KB PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238335
Brand: BINDER™ 80122371
Only null left
Add to basket
Add to basket
Non-adjustable analog output
  • Analog output
  • Not adjustable
  • For temperature values or temperature and humidity values
  • 4 to 20 mA
Specifications
For Use With (Equipment) KB 65/130/260/470/720/1060
Material Copper
Type Analog Output
Features For Temperature Values