missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Analytical Volumetric Sodium Hydroxide Solution, Reagecon™
Click to view available options
Quantity:
1 L
Description
his Analytical Volumetric Solution, also called titrant, standard titrant or standard solution, is certified to a specification of ± 0.1%. It is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete. The product may be used for manual titrations, or more commonly automatic titrations, to determine the end points by detection of any of several properties. Acid/base titrations, also called neutralisation titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.
Specifications
Specifications
Quantity | 1 L |
Packaging | HDPE Bottle GL45 |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'