missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

144.20 EUR - 410.97 EUR

Specifications

Antigen Angiopoietin 1
Concentration 0.31 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16802285
View Documents
Proteintech
23302-1-AP-20UL
20 μL
144.20 EUR
20µL
Please to purchase this item. Need a web account? Register with us today!
16892275
View Documents
Proteintech
23302-1-AP-150UL
150 μL
410.97 EUR
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
Specifications

Specifications

Angiopoietin 1
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Human, Rat, Mouse
O08538, O35460, Q15389
11600, 284, 89807
Angiopoietin 1 Fusion Protein Ag18829
Primary
-20°C
ANGPT1
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
AGP1, AGPT, ANG 1, ANG1, angiopoietin 1, ANGPT1, KIAA0003
ANGPT1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications