missing translation for 'onlineSavingsMsg'
Learn More

Scharlab ANILIN PA 250MLSegregation: Clear of living quarters. Separated f

Brand:  Scharlab AN03470250


ANILIN PA 250MLSegregation: Clear of living quarters. Separated f

Product Code. 11359853

  • ONLINE EXCLUSIVE PRICE
    147.00 EUR
    122.60 EUR / 250mL
    Save 24.40 EUR
    16% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aniline, reagent grade, ACS

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1

Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.