missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthraquinone-1-diazonium Chloride, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 16048-40-1 |
Molecular Formula | C14H7N2O2+ |
Quantity | 1 g |
Synonym | 1-anthracenediazonium, 9,10-dihydro-9,10-dioxo, 9,10-dihydro-9,10-dioxoanthracenediazonium, 1-aminoanthraquinone diazonium salt |
InChI Key | AYWYBCRYWKVVBR-UHFFFAOYSA-N |
SMILES | C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)[N+]#N |
IUPAC Name | 9,10-dioxoanthracene-1-diazonium |
Molecular Weight (g/mol) | 235.222 |
PubChem CID | 27658 |
Formula Weight | 16048-40-1 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction