missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 with control Mouse anti-Human, Unlabeled, Clone: 4D7, BD

Mouse Monoclonal Antibody

287.40 EUR - 858.00 EUR

Specifications

Antigen Bcl-2 with control
Clone 4D7
Applications Immunoassay, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15806528
View Documents
BD Biosciences
551097
50 μg
ONLINE EXCLUSIVE PRICE
288.00 EUR
287.40 EUR / Each
Save 0.60 EUR
0.21% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15816528
View Documents
BD Biosciences
551098
150 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The bcl-2 gene was first discovered in t(14:18) follicular lymphomas. Translocation of bcl-2 sequences from chromosome 18 onto the transcriptionally active immunoglobulin locus at chromosome band 14q32 in B-cells deregulates bcl-2 gene expression, resulting in high levels of bcl-2 mRNA. Bcl-2 protein blocks apoptosis (programmed cell death), and thereby may contribute to tomorigenesis by prolonging cell survival rather than by accelerating the rate of cell proliferation. Abundant evidence implicates this protein in the suppression of apoptosis in many types of cells. Bcl-2 is an intracellular membrane protein and resides primarily in the nuclear envelope, outer mitochondrial membrane and endoplasmic reticulum. The reduced molecular weight of Bcl-2 is 26 kDa. \parClone 4D7 recognizes human Bcl-2. A synthetic peptide containing amino acids 61-76 of the human Bcl-2 protein was used as immunogen. This peptide sequence differs from that of mouse Bcl-2 and hence 4D7 does not cross-react with mouse. 4D7 recognizes Bcl-2 as a 26 kDa band; however, additional higher molecular weight bands have been observed in some cell types. These bands may represent oligomers of Bcl-2, which have been reported to occur with members of the Bcl-2 family during the preparation of nonionic detergent extracts.

Specifications

Specifications

Bcl-2 with control
Immunoassay, Immunoprecipitation, Western Blot
Unconjugated
RUO
IgG1
Affinity Purified
4D7
Monoclonal
Mouse
Human
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.