Learn More
Invitrogen™ beta-1,2 Adaptin Monoclonal Antibody (100/1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA125065
Description
Recommended positive controls: rat brain, MDCK. This antibody stains clathrin-coated membranes at the plasma membrane, and in the Golgi region of methanol-fixed MDBK cells. By Western blot on bovine and rat brain preparations, this antibody detects a major band of AP-1 at ~115 kDa and a minor component at ~110 kDa, and bands at ~104 and ~112 kDa representing AP2. The antibody reacts with polypeptides at ~100 kDa in bovine liver, human heart fibroblasts and MDBK cells, but not with any components in the 110-115 kDa range from these sources, or from rat PC12 cells, neuroblastoma or astrocytes.
Adaptor protein complex 1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors. This complex is a heterotetramer composed of two large, one medium, and one small adaptin subunit. The protein encoded by this gene serves as one of the large subunits of this complex and is a member of the adaptin protein family. This gene is a candidate meningioma gene. Alternative splicing results in multiple transcript variants.
Specifications
| beta-1,2 Adaptin | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.1% sodium azide | |
| P52303, P62944, P63010, Q08DS7, Q10567 | |
| AP1B1, AP2B1 | |
| AP-2 adaptor protein from bovine brain. | |
| RUO | |
| 140670, 162, 163, 282183, 29663, 506192 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 100/1 | |
| Unconjugated | |
| AP2B1 | |
| adapter-related protein complex 1 subunit beta-1; adapter-related protein complex 2 beta subunit; adapter-related protein complex 2 subunit beta; adaptin, beta 2 (beta); adaptor protein complex AP-1 subunit beta-1; adaptor protein complex AP-1, beta 1 subunit; adaptor protein complex AP-2 subunit beta; adaptor related protein complex 1 beta 1 subunit; adaptor related protein complex 1 subunit beta 1; adaptor related protein complex 2 beta 1 subunit; adaptor related protein complex 2 subunit beta 1; adaptor related protein complex 2, beta 1 subunit; adaptor-related protein complex 1 beta 1 subunit; adaptor-related protein complex 1 subunit beta-1; adaptor-related protein complex 1, beta 1 subunit; adaptor-related protein complex 2 subunit beta; adaptor-related protein complex 2, beta 1 subunit; adaptor-related protein complex AP-1 beta 1 subunit; ADTB1; ADTB2; AP-1 complex subunit beta-1; AP105A; AP105B; AP1B1; AP-2 complex subunit beta; AP2B1; AP2-BETA; BAM22; beta adaptin; beta-1-adaptin; beta1-adaptin; Beta-2-adaptin; beta2-adaptin; beta-adaptin; Beta-adaptin 1; beta-prime-adaptin; Clapb1; CLAPB2; Clathrin assembly protein complex 1 beta large chain; clathrin assembly protein complex 2 beta large chain; clathrin-associated / assembly / adaptor protein, large, beta 1; clathrin-associated/assembly/adaptor protein, large, beta 1; golgi adaptor HA1/AP1 adaptin beta subunit; MGC128441 protein; plasma membrane adaptor HA2/AP2 adaptin beta subunit; plasma membrane adaptor HA2/AP2 adaptor beta subunit; testicular tissue protein Li 22 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Rat, Bovine | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.