missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Actin Loading Control, Biotin, clone: BA3R, Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen MA515739BTIN
This item is not returnable.
View return policy
Description
MA5-15739-BTIN has been successfully used for Western blot and FACS applications.
Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.
Specifications
| Beta-Actin Loading Control | |
| Monoclonal | |
| 1mg/mL | |
| ACTB | |
| Atcb | |
| Mouse | |
| Protein A | |
| RUO | |
| 100009272, 11461, 396526, 403580, 60, 81822 | |
| Store at 4°C. Do not freeze. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| BA3R | |
| Biotin | |
| O18840, P60706, P60709, P60710, P29751, P60711 | |
| ACTB | |
| Beta-actin N-terminal peptide. | |
| 50 μL | |
| Primary | |
| Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction