missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Collagen X Monoclonal Antibody (X-AC9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514268
This item is not returnable.
View return policy
Description
MA5-14268 targets Collagen Type X in IF and IHC (P, F) applications and shows reactivity with Chicken samples. The MA5-14268 immunogen is purified chicken type X collagen.
Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.
Specifications
| Collagen X | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P08125 | |
| COL10A1 | |
| Purified chicken type X collagen. | |
| 500 μL | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| X-AC9 | |
| Unconjugated | |
| COL10A1 | |
| Col10; COL10A1; Col10a-1; Collagen alpha-1(X) chain; collagen type X alpha 1 chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100858979 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction