Learn More
CPS1, Mouse, Clone: 4A10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CPS1.
Brand: Abnova H00001373-M02.100ug
Description
The protein encoded by this gene is an enzyme that catalyzes the first committed step of the hepatic urea cycle, which is important in the removal of excess urea from cells. There are two isozymes of this enzyme, and the encoded protein is the mitochondrial form. Three transcript variants encoding different isoforms have been found for this gene. The shortest isoform may not be localized to the mitochondrion. [provided by RefSeq
Sequence: ANNVPATPVAWPSQEGQNPSLSSIRKLIRDGSIDLVINLPNNNTKFVHDNYVIRRTAVDSGIPLLTNFQVTKLFAEAVQKSRKVDSKSLFHYRQYSAGKAASpecifications
| CPS1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001875 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1373 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 4A10 | |
| Mouse monoclonal antibody raised against a partial recombinant CPS1. | |
| CPS1 | |
| CPS1 | |
| CPS1 (NP_001866, 1400 a.a. ∼ 1500 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 λ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.