missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin C Mouse anti-Human, Clone: 197820, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB11962-100
This item is not returnable.
View return policy
Description
Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
Cystatin C | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 5-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P01034 | |
CST3 | |
Mouse myeloma cell line NS0-derived recombinant human Cystatin C Ser27-Ala146 Accession # P01034 | |
100 μg | |
Primary | |
Detects human Cystatin C in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human Cystatins A, B, D, E/M, F, S, SA, SN, X/Z/P, or recombinant mouse Cystatin C is observed. | |
Human | |
Supernatant |
Immunohistochemistry, ELISA | |
197820 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1471 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only