missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B Monoclonal Antibody (1F5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517071
This item is not returnable.
View return policy
Description
MA5-17071 targets EIF4B in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-17071 immunogen is purified recombinant fragment of human EIF4B expressed in E. Coli. MA5-17071 detects EIF4B which has a predicted molecular weight of approximately 80kDa.
The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.Specifications
eIF4B | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P23588, Q8BGD9 | |
Eif4b | |
Purified recombinant fragment of human EIF4B expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1F5 | |
Unconjugated | |
Eif4b | |
2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
Mouse | |
Protein G | |
RUO | |
1975, 75705 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction