Learn More
anti-GAPDH Matched Antibody Pair, Abnova™
Brand: Abnova H00002597-AP51.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-GAPDH, IgG1 κ (100μg); Detection antibody: rabbit purified polyclonal anti-GAPDH (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GAPDH
Host Species: Mouse, Rabbit
Isotype: IgG1 κ, IgG
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
GAPDH | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GAPDH. | |
G3PD/GAPD/MGC88685 | |
Mouse, Rabbit | |
RUO | |
2597 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
GAPDH | |
GAPDH | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-GAPDH, IgG1 Kappa (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-GAPDH (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
IgG, IgG1 κ |