Learn More
Mac-3 Rat anti-Mouse, Unlabeled, Clone: M3/84, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553322
345.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The M3/84 antibody reacts with the 110kDa Mac-3 antigen expressed on mouse mononuclear phagocytes. Surface expression of the 92 to 110kDa glycoprotein antigen increases during differentiation of monocytes to activated peritoneal macrophages. The M3/84 mAb can detect CD107b antigen on tissue macrophages, thioglycollate-elicited peritoneal macrophages, and some myeloid cell lines, but not on lymphocytes or monocytes. In the bone marrow, very few cells display CD107b antigen on the surface, but a large proportion express cytoplasmic CD107b. The M3/84 antibody has also been reported to stain dendritic cells and endothelium in sections of thymus (both medulla and cortex), lymph nodes, spleen (white pulp), and gut-associated lymphoid tissue.
Specifications
Mac-3 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse C57Bl/6 peritoneal exudate cells | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
M3/84 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.