Learn More
anti-MAPK3, MAPK1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0391.1Set
Additional Details : Weight : 0.00010kg
Includes: MAPK3 rabbit purified polyclonal antibody (20μg); MAPK1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAPK3 protein, and the other against the MAPK1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
MAPK3 | |
Monoclonal/Polyclonal | |
MAPK3 | |
MAPK3 | |
1 Set | |
Primary | |
Antibody pair set content: 1. MAPK3 rabbit purified polyclonal antibody (20 ug) 2. MAPK1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
ERK1/HS44KDAP/HUMKER1A/MGC20180/P44ERK1/P44MAPK/PRKM3 | |
Mouse, Rabbit | |
RUO | |
5595/5594 | |
Human | |
MAPK3 & MAPK1 Protein Protein Interaction Antibody Pair |