Learn More
VEGF Mouse anti-Human, Mouse, Rat, Porcine, Canine, Clone: VG1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 61 publication
Brand: Novus Biologicals NB100-664SS
Description
VEGF Monoclonal specifically detects VEGF in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| VEGF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry 1:20-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:100, Immunohistochemistry-Frozen 1:20-1:100, CyTOF-ready | |
| P15692 | |
| VEGFA | |
| Recombinant VEGF 189 protein. | |
| 0.025 mg | |
| Primary | |
| This VEGF Antibody (VG1) detects the 189, 165 and 121 isoforms of VEGF | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| VG1 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 7422 | |
| Human, Mouse, Rat, Porcine, Canine | |
| Purified |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only