missing translation for 'onlineSavingsMsg'
Learn More

p16INK4 with control Mouse anti-Human, Unlabeled, Clone: G175-405, BD

Mouse Monoclonal Antibody

182.49 EUR - 314.88 EUR

Specifications

Antigen p16INK4 with control
Clone G175-405
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15856558
View Documents
BD Biosciences
551153
50 μg
ONLINE EXCLUSIVE PRICE
183.00 EUR
182.49 EUR / Each
Save 0.51 EUR
0.28% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15866558
View Documents
BD Biosciences
551154
150 μg
ONLINE EXCLUSIVE PRICE
315.00 EUR
314.88 EUR / Each
Save 0.12 EUR
0.04% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Specifications

Specifications

p16INK4 with control
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
IgG1
Affinity Purified
G175-405
Monoclonal
Mouse
Human
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.