Learn More
p53 (acK382) Mouse anti-Human, Alexa Fluor 647, Clone: L82-51, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560231
Description
The p53 protein is critical to regulation of normal cell growth and proliferation and is a suppressor of tumor cell proliferation. Inactivation of p53 by a number of mechanisms, such as missense mutations or interaction with oncogenic viral or cellular proteins, can result in tumor progression. Mutations and/or allelic loss of the p53 gene are associated with a wide variety of human tumors. Known to have a role in transcriptional regulation, p53 suppresses various promoters containing TATA elements in an apparently sequence-independent fashion. p53 also binds to DNA in a sequence-specific manner via recognition of a 20-bp consensus-binding site. This interaction stimulates the expression of genes downstream of the p53 binding site. A number of genes that contain p53-binding sites have been identified, including MDM2, GADD45, and muscle creatine kinase. Post-translational acetylation of p53 enhances its DNA-binding activity. There are multiple factors that affect p53 acetylation, thereby modulating cellular proliferation and apoptosis.
The L82-51 monoclonal antibody recognizes acetylated lysine 382 (acK382) in the C-terminal region of p53.
Host Species: Mouse
Clone: L82-51
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human p53 acetylated Peptide
Intracellular Staining
Specifications
| p53 (acK382) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human p53 acetylated Peptide | |
| 50 Tests | |
| Cell Biology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| L82-51 | |
| TP53 (acK382) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.