missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PRDX5 Matched Antibody Pair, Abnova™
Brand: Abnova H00025824-AP22.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PRDX5 (100μg); Detection antibody: mouse purified polyclonal anti-PRDX5 (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PRDX5.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
PRDX5 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PRDX5. | |
ACR1/AOEB166/B166/MGC117264/MGC142283/MGC142285/PLP/PMP20/PRDX6/PRXV/SBBI10 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PRDX5 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-PRDX5 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
PRDX5 | |
PRDX5 | |
Affinity Purified | |
RUO | |
25824 | |
Human |