Learn More
Invitrogen™ SF3B3 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA518075
Product Code. 11366043
  • 520.00 EUR / 100µg
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
This gene encodes subunit 3 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex transcription coactivator-HAT complex, and the TFTC -containing complex). These complexes may function in chromatin modification, transcription, splicing, and DNA repair.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SF3B3
Polyclonal
Unconjugated
SF3B3
1810061H24Rik; 5730409A01Rik; AA409318; D8Ertd633e; Kiaa0017; mKIAA0017; pre-mRNA splicing factor SF3b, 130 kDa subunit; pre-mRNA-splicing factor SF3b 130 kDa subunit; RSE1; SAP 130; SAP130; SF3b130; SF3B3; Spliceosome-associated protein 130; splicing factor 3b subunit 3; splicing factor 3b, subunit 3; STAF130
Goat
Ammonium sulfate precipitation
RUO
23450
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q15393
SF3B3
Synthetic peptide sequence (VSKKLEDIRTRYAF) corresponding to the C-terminus amino acids of SF3B3 (aa 1204-1217).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions