Learn More
anti-TIMP1, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00007076-AP41.1Set
Product Code. 16162724
  • 809.00 EUR / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-TIMP1 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TIMP1, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
TIMP1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TIMP1.
CLGI/EPA/EPO/FLJ90373/HCI/TIMP
Mouse
RUO
7076
Human
Liquid
ELISA
Biotin
TIMP1
TIMP1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TIMP1 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TIMP1, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.