missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein B/ApoB Antibody (SR1898), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21714-100ul
This item is not returnable.
View return policy
Description
Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
| Apolipoprotein B/ApoB | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| SR1898 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 | |
| Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
| A synthesized peptide derived from human Apolipoprotein B/ApoB (Uniprot #: P04114) | |
| 100 μg | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 338 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction