Learn More
A&D Weighing Apollo Precision Balance Class II Approved
GREENER_CHOICE
Product Code. p-7203369
Click to view available options
Capacity:
220 g
320 g
420 g
620 g
2200 g
3200 g
4200 g
6200 g
Readability:
0.001 g
0.01 g
0.1 g
Unit Size:
Each
Product Code. 16864601
Brand: A&D Weighing GX203AEC
Apollo Analytical Series improves productivity and efficiency of your workflows with user-friendly and innovative features. It is designed to provide reliable results across a range of environments and applications.
  • Newly developed Smart SHS weighing sensor featuring ECL technology
  • APA (Auto Precision Assessment) – Automatic assessment of the balance performance, calculation and display of Standard Deviation (repeatability) at the point of use without the need for calibration weights, can be checked daily via a key press
  • Automatic Minimum Weight Calculation and setting via APA function
  • Input minimum weight automatically via APA or manually using weights
  • Visual Minimum Weight alert
  • UAC (User Access Control) – Administrator and Operator password access to ensure data integrity
  • Calibration via One-touch, Interval, pre-set times (3) or automatically in response to ambient temperature changes (GX-AE/A)
  • ISD (Patented Impact Shock Detection) – 4 level shock detection with visual and audible feedback, maintain the performance of your investment through correct use of the balance, ideal in automated loading applications and ideal for operator guidance
  • FRD (Flow Rate Display) – Direct display of flow rate of a sample in g/s, g/m, g/h or volume flow rate in mL/s, mL/m, mL/h
  • Up to 10 sample densities can be saved to memory
  • Use in conjunction with the flow Rate Comparator to keep flow rates within designated limits
  • SCF (Statistical Calculation Function) – Display and print various statistical results on the balance display or output to a printer or PC
  • Standard RS232 and Dual Mode USB interface. USB can be configured for quick mode (connect to PC without the need for software installation) or bi-directional mode
  • UFC (Universal Flexi Coms) – configurable data output, ideal for connection to data acquisition systems or barcode label printers etc.
  • Gross/Net/Tare data output, Tare memory, Underhook, Capacity Indicator
  • Large selection of Options and Accessories
  • Built-in fanless ionizer with Quick Ion™ technology and external IR switch (GX-AE)
Specifications
Certifications/Compliance GLP/GMP/GCP/ISO/OIML
Connection Type RS-232 C and USB
Capacity 220 g
Display Type Reverse-backlit LCD
Readability 0.001 g
Material Stainless Steel
Model GX-203A-EC
Stabilization Time 1 sec.
Length (Metric) Pan 128 mm
Width (English) Pan 5 in.
Width (Metric) Pan 128 mm
Electrical Requirements 30 VA to adapter (12 V, 1 A)