Learn More
STEMCELL Technologies ArciTect™ tracrRNA Kit
Product Code. p-7217422
Encompass_Preferred
Click to view available options
Quantity:
10 nmol Kit
20 nmol Kit
5 nmol Kit
Unit Size:
1 set
Product Code. 17109311
Brand: STEMCELL Technologies 76018
Trans-activating crRNA for guide RNA generation in CRISPR-Cas9 genome editing
  • ArciTect™ tracrRNA is a trans-activating CRISPR RNA (crRNA), one of two RNA components required to make a guide RNA (gRNA) for CRISPR-Cas9 genome editing
  • ArciTect™ tracrRNA contains a complementary sequence to the linker sequence in ArciTect™ crRNA, allowing fast and efficient gRNA duplex formation
  • It is recognized by all ArciTect™ Cas9 nucleases and is essential for ribonucleoprotein (RNP) complex formation
  • It contains 2'-O-methyl and phosphorothioate modifications at the first two 5' and 3' terminal residues for optimal stability and editing efficiency
  • The ArciTect™ tracrRNA Kit includes tracrRNA as well as ArciTect™ Annealing Buffer (5X), which is required for forming a gRNA duplex between ArciTect™ tracrRNA and ArciTect™ crRNA
  • ArciTect™ crRNA is available by custom order for user-specified target sequences, in 2 nmol (Catalog #76010), 10 nmol (Catalog #76011), and 20 nmol (Catalog #76012) sizes
  • ArciTect™ Cas9 Nuclease is also available separately (Catalog #76001)
Specifications
Cell Type Pluripotent Stem Cells
Quantity 20 nmol Kit