missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11112-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a Rho guanine nucleotide exchange factor (GEF). Rho GEFs regulate the activity of small Rho GTPases by stimulating the exchange of guanine diphosphate (GDP) for guanine triphosphate (GTP) and may play a role in neural morphogenesis. Mutations in this gene are associated with slowed nerve conduction velocity (SNCV). Alternative splicing results in multiple transcript variants.Specifications
ARHGEF10 | |
Polyclonal | |
Unconjugated | |
ARHGEF10 | |
ARHGEF10, GEF10, KIAA0294 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234094, 306618, 9639 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15013, Q8C033 | |
ARHGEF10 | |
ARHGEF10 Fusion Protein Ag1605 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |