missing translation for 'onlineSavingsMsg'
Learn More

ATF3 Rabbit anti-Human, Clone: S08-6H6, Novus Biologicals™

Rabbit Monoclonal Antibody

285.09 EUR - 577.74 EUR

Specifications

Antigen ATF3
Clone S08-6H6
Dilution Western Blot, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18331404
View Documents Promotion Details
Novus Biologicals
NBP3-15001-25UL
25 μg
285.09 EUR
25µL
Please to purchase this item. Need a web account? Register with us today!
18389295
View Documents Promotion Details
Novus Biologicals
NBP3-15001-100UL
100 μg
577.74 EUR
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATF3 Monoclonal antibody specifically detects ATF3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

ATF3
Western Blot, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Purified
RUO
Human
activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3
Recombinant protein of human ATF3 (Uniprot#: P18847)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
S08-6H6
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Transcription Factors and Regulators
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
467
IgG
Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications