missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Atherogenic Diet

Semi-purified atherogenic diet modified for mice.

668.00 EUR - 2515.00 EUR

Specifications

Product Type Animal Diet
For Use With (Application) Rodent diet
Storage Requirements 2°C to 8°C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11471452
View Documents
MP Biomedicals™
0296040410
10 kg
668.00 EUR
10kg
Please to purchase this item. Need a web account? Register with us today!
11481452
View Documents
MP Biomedicals™
0296040420
20 kg
1140.00 EUR
20kg
Please to purchase this item. Need a web account? Register with us today!
11491452
View Documents
MP Biomedicals™
0296040450
50 kg
2515.00 EUR
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Atherogenic diet is rich in cholesterol and other atherogenic factors
  • Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease
Specifications

Specifications

Animal Diet
Rodent diet
2°C to 8°C
Videos
SDS
Documents

Documents

Product Certifications

Atherogenic diet is rich in cholesterol and other atherogenic factors. Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease.