missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Synthase B1 Polyclonal Antibody

ATP Synthase B1 Polyclonal Antibody
Brand: Invitrogen PA5100034
This item is not returnable.
View return policy
Description
ATP Synthase B1 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| ATP Synthase B1 | |
| Polyclonal | |
| Unconjugated | |
| ATP5PB | |
| ATP synthase B chain, mitochondrial; ATP synthase F(0) complex subunit B1, mitochondrial; ATP synthase peripheral stalk-membrane subunit b; ATP synthase proton-transporting mitochondrial F(0) complex subunit B1; ATP synthase subunit b; ATP synthase subunit b, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1; Atp5f; ATP5F1; Atp5pb; ATPase subunit b; C76477; cell proliferation-inducing protein 47; H+-ATP synthase subunit b; PIG47 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11950, 171375, 515 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19511, P24539, Q9CQQ7 | |
| ATP5PB | |
| A synthesized peptide derived from human ATP5F1(Accession P24539), corresponding to amino acid residues P43-Y93. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction