missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPA, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A selective and potent GluK1 (formerly GluR5) kainate receptor agonist (Ki = 4.3 nM), inactive at GluK6 (formerly GluR6) (Ki > 1 mM) and only weakly active at AMPA receptors (GluA1-4) and the kainate receptors GluK5 (formerly KA-2) and GluK3 (formerly GluR7) (Ki values of 6 - 14 μM).
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 228.25 |
Percent Purity | >98% |
Chemical Name or Material | ATPA |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'