missing translation for 'onlineSavingsMsg'
Learn More

ATPA, Tocris Bioscience™

Code produit. 30001387
Click to view available options
Quantity:
10 mg
50 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Potent and selective GluK1 kainate agonist

A selective and potent GluK1 (formerly GluR5) kainate receptor agonist (Ki = 4.3 nM), inactive at GluK6 (formerly GluR6) (Ki > 1 mM) and only weakly active at AMPA receptors (GluA1-4) and the kainate receptors GluK5 (formerly KA-2) and GluK3 (formerly GluR7) (Ki values of 6 - 14 μM).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Identifiants chimiques

CAS 140158-50-5
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.248
InChI Key PIXJURSCCVBKRF-UHFFFAOYSA-N
Synonym atpa, atpa-tert, r,s-atpa, alpha-amino-3-hydroxy-5-tert-butyl-4-isoxazolepropionate, 2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propionic acid, rs-2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo-4-isoxazolepropanoic acid, 4-isoxazolepropanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo, atpa, solid, acmc-20cirp
PubChem CID 2253
IUPAC Name 2-amino-3-(5-tert-butyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
SMILES CC(C)(C)C1=C(C(=O)NO1)CC(C(=O)O)N

Spécification

Quantity 50 mg
Formula Weight 228.25
Percent Purity >98%
Chemical Name or Material ATPA
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'