Learn More
Invitrogen™ Attune™ NxT Focusing Fluid Filter
This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune™ NxT Acoustic Focusing Cytometer.
Brand: Invitrogen™ 100022587
This item is not returnable.
View return policy
Description
This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune™ NxT Acoustic Focusing Cytometer. Filters are recommended for replacement once every three months, unless performing small particle detection when once-a-month replacement is recommended.
Specifications
Attune™ | |
1 Filter | |
Focusing Fluid Filter |
Store at room temperature. | |
Attune Acoustic Focusing Cytometer |