Learn More
Abeba™ Autoclavable Clogs 39620
Product Code. 15611268
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. 15611268
Brand: Abeba™ 39620/45
Antistatic and autoclavable at 135°C for 5 minutes. High slip resistance. Washing at 60°C.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color Green
Size 45
Sole Features Slip-resistant
Style 39620