missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

Product Code. 11320135 Shop All Axygen Scientific Products
Click to view available options
Color:
Blue
Clear
Sterility:
Non-sterile
Sterile
No. of Wells:
24
48
96
384
Volume (Metric) Well:
1.1 mL
1.8 mL
10,000 μL
1100 μL
125 μL
2000 μL
240 μL
5000 μL
550 μL
Unit Size:
Pack of 10
Pack of 25
Pack of 30
Pack of 5
Pack of 50
This item is not returnable. View return policy

Product Code. 11320135

Brand: Axygen™ PDW11CS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For sample collection, in-vitro growth chambers and long term storage.

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SLAS 1-2004
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Autoclavable Autoclavable
    Color Clear
    Product Type 96 Round Deep-Well Storage Microplate
    Material Polypropylene
    Volume (Metric) Well 1100 μL
    No. of Wells 96
    Sterility Sterile
    Well Shape Round
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Max. RCF 4000 x g
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.