Learn More
Azlon™ HDPE Measuring Scoop
Sturdy yet lightweight polyethylene scoop. The Azlon™ HDPE measuring scoop is chemical resistant and suitable for food contact.
Brand: Azlon SWL028
28.98 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.17550kg
Description
- Sturdy scoops in natural rigid polyethylene
- Lightweight with good chemical resistance
- Food safe
Specifications
Measuring Scoop | |
HDPE | |
150 mL | |
250 mm |