Learn More
B-Myb Antibody (1C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00004605-M02
Product Code. 18311568
  • 589.00 EUR / 0.01mg
Description
B-Myb Monoclonal antibody specifically detects B-Myb in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B-Myb
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
B-MYB, BMYBMGC15600, Myb-like protein 2, myb-related protein B, v-myb avian myeloblastosis viral oncogene homolog-like 2, v-myb myeloblastosis viral oncogene homolog (avian)-like 2
MYBL2 (AAH53555, 601 a.a. ~ 700 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TLPKSLSLPTTAPSNSSSLTLSGIKEDNSLLNQGFLQAKPEKAAVAQKPRSHFTTPAPMSSAWKTVACGGTRDQLFMQEKARQLLGRLKPSHTSRTLILS
0.1 mg
Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers
4605
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
1C7
Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
AAH53555
Mouse
IgG purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions