Learn More
Masterflex™ B/T™ Analog Drive
Masterflex™ B/T™ Analog Drive for use with high volume transfer and dispensing
Brand: Masterflex™ 77111-37
This item is not returnable.
View return policy
Description
- Masterflex™ B/T™ drives are the best choice for high-flow bulk-transfer applications
- Their durable construction ensures long service life with minimal maintenance
- The sturdy materials of construction include stainless steel/carbon steel, a cast aluminum housing with an epoxy-powder coating, and Nylatron™ rollers for rugged dependability
- All B/T™ drives include a new and improved Rapid-Load™ pump head for fast and easy tubing changes
- The redesigned occlusion mechanism provides an improved tubing retention mechanism which holds the tubing firmly in place to further simplify the loading process
- PerfectPosition™ tubing sizes provide improved performance in higher-pressure applications and provide longer tubing life
Specifications
6.3 amps | |
(38.4 x 68.6 x 40 cm) | |
16 to 35 LPM | |
1 h/p | |
B/T™ | |
Wash-Down Pump |
Fixed-Speed | |
220 VAC | |
IP55-rated | |
3 | |
B/T 87, or B/T 91 | |
≤271 rpm |