Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels
Nylon cloth labels for BBP33/i3300 printers.
Brand: Brady™ 361682
This item is not returnable.
View return policy
Description
For BBP33/i3300 printers.
Specifications
Permanent Acrylic | |
Matte | |
Labels | |
Rectangle | |
66.68 mm | |
25.40 mm |
White | |
Nylon Cloth (B-499) | |
Non-sterile | |
25.40 mm (W) x 66.68 mm (H) | |
25.40 mm |