Learn More
CPC™ Back-to-Back Body Adapter
Product Code. 17425103
Click to view available options
:
Polysulfone, USP Class VI, ADCF, sterlizations: Gamma Autoclave, free floating, -40 F to 300 F
Product Code. 17425103
Brand: CPC™ MPC17X1739
Only null left
Add to basket
Add to basket
MPC Body x MPX Back-to-Back Body Adapter
  • Medical Grade
  • Non-valved
  • Natural finish
Specifications
Certifications/Compliance USP Class VI, ADCF
Material Polysulfone
Min. Operating Temperature (English) -40°F
Sterilization Method Gamma, Autoclave
Color Amber
Maximum Pressure 60 psi, 4.1 bar
Max. Operating Temperature (English) 300°F
Max. Operating Temperature (Metric) 149°C