Learn More
Masterflex™ BarbLock Assemblies
Click to view available options
For Use With:
Fits 1/4 in. and L/S 15, 17, & 24 Tubing
Fits 1/4 in. x 3/8 in. Tubing
Unit Size:
Each
Description
- Safe to use with pressures up to 140psi or upper pressure limit of tubing
- An excellent alternative to securing tubing and hose barbs with cable ties — no more crimps or tube distortion
- BarbLock assembly consists of three pieces: the sleeve, collect, and barbed fittings
- To install the fitting, slide the sleeve and collet onto the tubing and then inserts the barbed fitting into the tubing
- Prelock the sleeve and collet by pressing them together until you hear a click
- Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool
- Once assembled, the BarbLock assembly can only be unlocked with the removal tool
Specifications
Specifications
| For Use With | Fits 1/4 in. x 3/8 in. Tubing |
| For Use With (Application) | Easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage |
| Packaging Quantity | 1 Pack |
| Product Type | BarbLock Assembly |