Learn More
Saint Gobain Life Sciences BarbLock™ Removal Tool
Universal removal tool.
136.22 EUR - 243.72 EUR
Product Code | Brand | Product Line | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Line | Price | Quantity & Availability | |||||
30040239
|
Saint Gobain Life Sciences
BLTURT |
500 Series |
136.22 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30040267
|
Saint Gobain Life Sciences
BLTURT II |
625 Series |
243.72 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
- Constructed from high-performance polypropylene that meets USP Class VI criteria
- Quick and easy to assemble
- Compatible with all flexible tubing material
- Disposable assemblies
- Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
- Autoclavable
Specifications
Tubing Clamp |