Learn More
Invitrogen™ Barrier (Filter) Tips, 1,000 μL size
Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are RNase- and DNase-free and are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals.
Brand:  Invitrogen™ AM12665TS
176.70 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 2.58500kg
Product Code. 17779347
  • 186.00 EUR / Each
Estimated Shipment: 12-07-2024
Description
  • Ambion Barrier tips prevent cross-contamination of samples by means of an aerosol filter
  • They are RNase- and DNase-free and are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals
  • Ten racks containing 96 tips each are provided
  • Tips are compatible with Eppendorf, Finnpipette, Oxford Benchmate, Pipetman and other brands of pipettors
  • Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
Specifications
Specifications
1000 μL
Sterile
Barrier
Room Temperature
Invitrogen
Filtered
Rack
Includes 10 racks with 96 tips each.
Store at room temperature.
Universal Fit