missing translation for 'onlineSavingsMsg'
Learn More

Batefenterol, MedChemExpress

Product Code. 16461637
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461637

Brand: Medchem Express HY12980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Batefenterol (GSK961081;TD-5959) is a novel muscarinic receptor antagonist and β2-adrenoceptor agonist; displays high affinity for hM2, hM3 muscarinic and hβ2-adrenoceptor with Ki values of 1.4, 1.3 and 3.7 nM, respectively.

Batefenterol (GSK961081

Specifications

Chemical Name or Material Batefenterol
CAS 743461-65-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C40H42ClN5O7
Quantity 50 mg
Synonym GSK961081 TD-5959
Solubility Information DMSO : ≥ 100 mg/mL (135.09 mM)
SMILES O=C(OC1CCN(CCC(NC2=CC(OC)=C(CNC[C@H](O)C3=CC=C(O)C4=C3C=CC(N4)=O)C=C2Cl)=O)CC1)NC5=CC=CC=C5C6=CC=CC=C6
Molecular Weight (g/mol) 740.24
Formula Weight 740.24
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.