missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Baume Hydrometer, Heavy Liquids

The Baume (Heavy) Hydrometer is for liquids heavier than water.

Brand:  VEE GEE 6609-14

 View more versions of this product

Product Code. 17846443

  • 33.60 EUR / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Specific Gravity: 145/(145-degrees Baume)
  • Calibrated for use at 15.5°C/60°F
Specifications

Specifications

Baume Hydrometer
30.5 cm
0 to 70°
1.0°
Glass
15.5°C (60°F)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers