missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bax inhibitor 1 Antibody (20F565), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
236.00 EUR - 477.00 EUR
Specifications
| Antigen | Bax inhibitor 1 |
|---|---|
| Clone | 20F565 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry-Paraffin 0.5-5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18174734
|
Novus Biologicals
NBP2-24912SS |
0.025 mg |
236.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726133
|
Novus Biologicals
NBP2-24912 |
477.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Bax inhibitor 1 Monoclonal specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Bax inhibitor 1 | |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| P55061 | |
| 7009 | |
| A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40) UniProt P55061. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 20F565 | |
| Western Blot 1-5 ug/ml, Immunohistochemistry-Paraffin 0.5-5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6 | |
| TMBIM6 | |
| IgG2b κ | |
| Protein G purified | |
| This antibody is reactive to Human, Mouse, Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title