Learn More
BD BBL™ Taxo™ Blank Discs
Provides a cost effective identification and differentiation tool. BD™ BBL™ Taxo™ Blank Discs are easy-to-use discs for classical identification of microorganisms. The Taxo product line includes discs for identifying a variety of organisms.
128.00 EUR
Specifications
For Use With (Application) | Identification and differentiation of microorganisms |
---|---|
For Use With (Equipment) | Taxo™ brand testing disc |
Packaging Type | BBL Blank Test Discs |
Quantity | 6 Pack |
Description
Order Info
50 discs per vial; six vials per pack
Specifications
Identification and differentiation of microorganisms | |
Taxo™ brand testing disc | |
BBL Blank Test Discs | |
6 Pack |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days